3,6-Dibromo-9-(triphenylen-2-yl)carbazole ≥98.0% (by HPLC)
Fornitore: TCI EUROPE
|
Attenzione
|
D4910-200MG
D4910-1G
TCIAD4910-200MGEA
103
EUR
TCIAD4910-200MG
TCIAD4910-1G
3,6-Dibromo-9-(triphenylen-2-yl)carbazole ≥98.0% (by HPLC)
3,6-Dibromo-9-(triphenylen-2-yl)carbazole
Formula:
C₃₀H₁₇Br₂N Peso molecolare: 551,28 g/mol |
CAS Number:
1351870-16-0 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...