Bis(α,α-dimethylbenzyl)peroxide (dicumyl peroxide) (contains ca. 60% CaCO3)
Fornitore: TCI
|
Pericolo
|
Synonyms:
Bis(1-methyl-1-phenylethyl)peroxide, Bis(alpha,alpha-dimethylbenzyl)peroxide, Dicumyl Peroxide
Formula:
C₁₈H₂₂O₂ Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00036227 CAS Number: 80-43-3 EINECS: 201-279-3 UN: 3077 ADR: 9,III |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Calcium carbonate | 57.0 to 60.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...