Stampa…

2,5-Dihydroxybenzoic acid sodium salt hydrate ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: Gentisic acid sodium salt

D4893-5G D4893-25G
TCIAD4893-5GEA 36 EUR
TCIAD4893-5G TCIAD4893-25G
2,5-Dihydroxybenzoic acid sodium salt hydrate ≥98.0% (by HPLC, titration analysis)
2,5-Dihydroxybenzoic acid sodium salt hydrate
Formula: C₇H₅NaO₄·xH₂O
Peso molecolare: 176,10 g/mol
MDL Number: MFCD00150273
CAS Number: 4955-90-2
EINECS: 225-598-2
Merck Index: 14,04398

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Water 10.0 to 20.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR