Stampa…

2,4-Dichloro-1,3,5-triazine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

D4891-1G D4891-5G
TCIAD4891-1GEA 56 EUR
TCIAD4891-1G TCIAD4891-5G
2,4-Dichloro-1,3,5-triazine ≥98.0% (by GC, titration analysis)
2,4-Dichloro-1,3,5-triazine
Formula: C₃HCl₂N₃
Peso molecolare: 149,9661 g/mol
Punto di ebollizione: 102°C
MDL Number: MFCD04115347
CAS Number: 2831-66-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 54.0 to 58.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR