Stampa…

(S)-(-)-6,6'-Dibromo-2,2'-bis(methoxymethoxy)-1,1'-binaphthyl ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: (S)-6,6'-Dibromo-2,2'-bis(methoxymethoxy)-1,1'-binaphthalene, (S)-6,6'-Dibromo-2,2'-bis(methoxymethoxy)-1,1'-binaphthyl

D4883-1G D4883-5G
TCIAD4883-1GEA 89 EUR
TCIAD4883-1G TCIAD4883-5G
(S)-(-)-6,6'-Dibromo-2,2'-bis(methoxymethoxy)-1,1'-binaphthyl ≥98.0% (by HPLC, titration analysis)
(S)-(-)-6,6'-Dibromo-2,2'-bis(methoxymethoxy)-1,1'-binaphthyl
Formula: C₂₄H₂₀Br₂O₄
Peso molecolare: 532,23 g/mol
MDL Number: MFCD03788936
CAS Number: 211560-97-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0 %
Melting point 132.0 to 136.0 deg-C
Specific rotation [a]20/D -20.0 to -23.0 deg (C=1,CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR