Stampa…

4-Oxo-1,4-dihydroquinoline-3-carboxylic acid ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI
Attenzione

D4881-1G D4881-5G
TCIAD4881-1GEA 29 EUR
TCIAD4881-1G TCIAD4881-5G
4-Oxo-1,4-dihydroquinoline-3-carboxylic acid ≥98.0% (by HPLC, titration analysis)
4-Oxo-1,4-dihydroquinoline-3-carboxylic acid
Formula: C₁₀H₇NO₃
MDL Number: MFCD00498984
CAS Number: 13721-01-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR