Stampa…

2,8-Diiododibenzothiophene ≥97.0% (by GC)

Fornitore: TCI EUROPE

D4870-200MG D4870-1G
TCIAD4870-200MGEA 70 EUR
TCIAD4870-200MG TCIAD4870-1G
2,8-Diiododibenzothiophene ≥97.0% (by GC)
2,8-Diiododibenzothiophene
Formula: C₁₂H₆I₂S
Peso molecolare: 436,05 g/mol
MDL Number: MFCD00275724
CAS Number: 105404-91-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 223.0 to 227.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR