4,6-Dibromodibenzofuran ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
4,6-Dibromodibenzo[b,d]furan
D4858-200MG
D4858-1G
TCIAD4858-200MGEA
28
EUR
TCIAD4858-200MG
TCIAD4858-1G
4,6-Dibromodibenzofuran ≥98.0% (by GC)
4,6-Dibromodibenzofuran
Formula:
C₁₂H₆Br₂O Peso molecolare: 325,99 g/mol |
MDL Number:
MFCD00185686 CAS Number: 201138-91-2 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 147.0 to 151.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...