Stampa…

4-Diethylaminobenzaldoxime ≥95.0% (by GC)

Fornitore: TCI
Attenzione

Synonyms: 4-Diethylaminobenzaldehyde Oxime, 4-Diethylaminobenzaldoxime (mixture of isomers)

D4850-1G D4850-5G
TCIAD4850-1GEA 23 EUR
TCIAD4850-1G TCIAD4850-5G
4-Diethylaminobenzaldoxime ≥95.0% (by GC)
4-Diethylaminobenzaldoxime
Formula: C₁₁H₁₆N₂O
MDL Number: MFCD00051801
CAS Number: 54376-65-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 95.0 %
Melting point 93.0 to 97.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR