2,4-Diethoxybenzaldehyde ≥98.0% (by GC)
Fornitore: TCI
Formula:
C₁₁H₁₄O₃ |
MDL Number:
MFCD00995010 CAS Number: 22924-16-9 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | gt. 70.0 to 74.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...