Stampa…

4,4'-Dichlorochalcone ≥98.0% (by GC)

Fornitore: TCI
Attenzione

D4846-5G D4846-25G
TCIAD4846-5GEA 65 EUR
TCIAD4846-5G TCIAD4846-25G
4,4'-Dichlorochalcone ≥98.0% (by GC)
4,4'-Dichlorochalcone
Formula: C₁₅H₁₀Cl₂O
MDL Number: MFCD00018704
CAS Number: 19672-59-4

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 156.0 to 160.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR