Stampa…

N,N-Diphenyl-2-naphthylamine ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: N,N-Diphenylnaphthalen-2-amine

D4843-1G D4843-5G
TCIAD4843-1GEA 74 EUR
TCIAD4843-1G TCIAD4843-5G
N,N-Diphenyl-2-naphthylamine ≥98.0% (by GC)
N,N-Diphenyl-2-naphthylamine
Formula: C₂₂H₁₇N
Peso molecolare: 295,39 g/mol
MDL Number: MFCD28384144
CAS Number: 6940-30-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 123.0 to 127.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR