7,8-Dimethoxy-1,3-dihydro-2H-3-benzazepin-2-one ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
7,8-Dimethoxy-1H-benzo[d]azepin-2(3H)-one, 1,3-Dihydro-7,8-dimethoxy-2H-3-benzazepin-2-one
D4824-1G
D4824-5G
TCIAD4824-1GEA
49
EUR
TCIAD4824-1G
TCIAD4824-5G
7,8-Dimethoxy-1,3-dihydro-2H-3-benzazepin-2-one ≥98.0% (by GC)
7,8-Dimethoxy-1,3-dihydro-2H-3-benzazepin-2-one
Formula:
C₁₂H₁₃NO₃ Peso molecolare: 219,2400 g/mol |
MDL Number:
MFCD02091565 CAS Number: 73942-87-7 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 244.0 to 248.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...