Stampa…

2-(2,4-Dihydroxyphenyl)-4,6-diphenyl-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE
Attenzione

D4819-1G D4819-5G
TCIAD4819-1GEA 53 EUR
TCIAD4819-1G TCIAD4819-5G
2-(2,4-Dihydroxyphenyl)-4,6-diphenyl-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)
2-(2,4-Dihydroxyphenyl)-4,6-diphenyl-1,3,5-triazine
Formula: C₂₁H₁₅N₃O₂
Peso molecolare: 341,37 g/mol
Punto di fusione: 276°C
MDL Number: MFCD16251126
CAS Number: 38369-95-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR