Stampa…

1-(2,4-Dihydroxyphenyl)-2-(4-methoxyphenyl)ethanone ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

Synonyms: 2,4-Dihydroxyphenyl 4-Methoxybenzyl ketone, 2',4'-Dihydroxy-2-(4-methoxyphenyl)acetophenone

D4816-200MG D4816-1G
TCIAD4816-200MGEA 28 EUR
TCIAD4816-200MG TCIAD4816-1G
1-(2,4-Dihydroxyphenyl)-2-(4-methoxyphenyl)ethanone ≥98.0% (by titrimetric analysis)
1-(2,4-Dihydroxyphenyl)-2-(4-methoxyphenyl)ethanone
Formula: C₁₅H₁₄O₄
Peso molecolare: 258,28 g/mol
MDL Number: MFCD00020119
CAS Number: 487-49-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 158.0 to 162.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR