Stampa…

2,4-Dichloro-6-morpholino-1,3,5-triazine ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-(4,6-Dichloro-1,3,5-triazin-2-yl)morpholine, 2,4-Dichloro-6-(4-morpholinyl)-1,3,5-triazine

D4814-1G D4814-5G
TCIAD4814-1GEA 38 EUR
TCIAD4814-1G TCIAD4814-5G
2,4-Dichloro-6-morpholino-1,3,5-triazine ≥98.0% (by GC)
2,4-Dichloro-6-morpholino-1,3,5-triazine
Formula: C₇H₈Cl₂N₄O
Peso molecolare: 235,0706 g/mol
MDL Number: MFCD00460178
CAS Number: 6601-22-5
EINECS: 229-544-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 159.0 to 163.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR