Stampa…

4,5-Dimethoxy-2-nitrobenzyl alcohol ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 6-Nitroveratryl alcohol, (4,5-Dimethoxy-2-nitrophenyl)methanol

D4811-1G D4811-5G
TCIAD4811-1GEA 45 EUR
TCIAD4811-1G TCIAD4811-5G
4,5-Dimethoxy-2-nitrobenzyl alcohol ≥98.0% (by GC)
4,5-Dimethoxy-2-nitrobenzyl alcohol
Formula: C₉H₁₁NO₅
Peso molecolare: 213,19 g/mol
MDL Number: MFCD00014701
CAS Number: 1016-58-6
EINECS: 213-806-4

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 147.0 to 151.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR