Stampa…

3,5-Dibromo-α,α,α-trifluorotoluene ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3,5-Dibromobenzotrifluoride, 1,3-Dibromo-5-(trifluoromethyl)benzene

D4807-5G D4807-25G
TCIAD4807-5GEA 41 EUR
TCIAD4807-5G TCIAD4807-25G
3,5-Dibromo-α,α,α-trifluorotoluene ≥98.0% (by GC)
3,5-Dibromo-α,α,α-trifluorotoluene
Formula: C₇H₃Br₂F₃
Peso molecolare: 303,90 g/mol
MDL Number: MFCD00526219
CAS Number: 401-84-3

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR