Stampa…

δ-methrin ≥97.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: (S)-alpha-Cyano-3-phenoxybenzyl (1R,3R)-3-(2,2-Dibromovinyl)-2,2-dimethylcyclopropanecarboxylate, (1R,3R)-3-(2,2-Dibromovinyl)-2,2-dimethylcyclopropanecarboxylic acid (S)-alpha-Cyano-3-phenoxybenzyl ester, Deltamethrin

D4775-100MG D4775-1G
TCIAD4775-100MGEA 35 EUR
TCIAD4775-100MG TCIAD4775-1G
δ-methrin ≥97.0% (by GC)
Deltamethrin
Formula: C₂₂H₁₉Br₂NO₃
Peso molecolare: 505,206 g/mol
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00870122
CAS Number: 52918-63-5
EINECS: 258-256-6
UN: 2811
ADR: 6.1,II
Merck Index: 14,02883

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 97.0 %
Melting point 99.0 to 103.0 deg-C
Specific rotation [a]20/D +22.0 to +26.0 deg (C=1, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR