Stampa…

4-(Difluoromethoxy)benzoic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

D4762-1G D4762-5G
TCIAD4762-1GEA 70 EUR
TCIAD4762-1G TCIAD4762-5G
4-(Difluoromethoxy)benzoic acid ≥98.0% (by GC, titration analysis)
4-(Difluoromethoxy)benzoic acid
Formula: C₈H₆F₂O₃
MDL Number: MFCD00221454
CAS Number: 4837-20-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 168.0 to 172.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR