Stampa…

4'-(Difluoromethoxy)acetophenone ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1-(4-(Difluoromethoxy)phenyl)ethanone

D4760-1G D4760-5G
TCIAD4760-1GEA 67 EUR
TCIAD4760-1G TCIAD4760-5G
4'-(Difluoromethoxy)acetophenone ≥98.0% (by GC)
4'-(Difluoromethoxy)acetophenone
Formula: C₉H₈F₂O₂
Peso molecolare: 186,1600 g/mol
MDL Number: MFCD00042250
CAS Number: 83882-67-1
EINECS: 281-174-7

Visualizza i codici

Risultati del test di specificazione

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR