Stampa…

2,5-Dichloro-3,4-ethylenedioxythiophene ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 5,7-Dichloro-2,3-dihydrothieno[3,4-b][1,4]dioxine

D4746-1G D4746-5G
TCIAD4746-1GEA 73 EUR
TCIAD4746-1G TCIAD4746-5G
2,5-Dichloro-3,4-ethylenedioxythiophene ≥98.0% (by GC)
2,5-Dichloro-3,4-ethylenedioxythiophene
Formula: C₆H₄Cl₂O₂S
Peso molecolare: 211,06 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD09907757
CAS Number: 225518-49-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 65.0 to 69.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR