Stampa…

5,6-Dibromo-1,3-benzodioxole ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 5,6-Dibromobenzo[d][1,3]dioxole, 1,2-Dibromo-4,5-(methylenedioxy)benzene

D4737-1G D4737-5G
TCIAD4737-1GEA 19 EUR
TCIAD4737-1G TCIAD4737-5G
5,6-Dibromo-1,3-benzodioxole ≥98.0% (by GC)
5,6-Dibromo-1,3-benzodioxole
Formula: C₇H₄Br₂O₂
Peso molecolare: 279,93 g/mol
MDL Number: MFCD00094476
CAS Number: 5279-32-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 84.0 to 88.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR