2,6-Diiodo-4-nitroaniline ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
D4736-5G
D4736-25G
TCIAD4736-5GEA
90
EUR
TCIAD4736-5G
TCIAD4736-25G
2,6-Diiodo-4-nitroaniline ≥98.0% (by GC)
2,6-Diiodo-4-nitroaniline
Formula:
C₆H₄I₂N₂O₂ Peso molecolare: 389,92 g/mol |
MDL Number:
MFCD00007683 CAS Number: 5398-27-6 EINECS: 226-429-5 |
Risultati del test di specificazione
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 254.0 to 258.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...