Di-tert-butyl(methyl)phosphonium tetrafluoroborate ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
(t-Bu)2PMeHBF4
D4731-1G
D4731-5G
TCIAD4731-1GEA
113
EUR
TCIAD4731-1G
TCIAD4731-5G
Di-tert-butyl(methyl)phosphonium tetrafluoroborate ≥98.0% (by titrimetric analysis)
Di-tert-butyl(methyl)phosphonium tetrafluoroborate
Formula:
C₉H₂₂BF₄P Peso molecolare: 248,05 g/mol |
MDL Number:
MFCD03840579 CAS Number: 479094-62-7 UN: 1759 ADR: 8,III |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Iodometric Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...