2,6-Dichloro-3-pyridineboronic acid (contains varying amounts of Anhydride)
Fornitore: TCI
|
Attenzione
|
Synonyms:
2,6-Dichloropyridin-3-ylboronic acid, 2,6-Dichloropyridine-3-boronic acid
Formula:
C₅H₄BCl₂NO₂ Punto di fusione: 150°C |
MDL Number:
MFCD03094989 CAS Number: 148493-34-9 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 110.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...