Stampa…

N-(2-Butyl-3-(4-(3-(dibutylamino)propoxy)benzoyl)benzofuran-5-yl)methanesulphonamide hydrochloride ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Dronedarone hydrochloride, N-[2-Butyl-3-[4-[3-(dibutylamino)propoxy]benzoyl]-5-benzofuranyl]methanesulphonamide hydrochloride

D4689-200MG D4689-1G
TCIAD4689-200MGEA 39 EUR
TCIAD4689-200MG TCIAD4689-1G
N-(2-Butyl-3-(4-(3-(dibutylamino)propoxy)benzoyl)benzofuran-5-yl)methanesulphonamide hydrochloride ≥98.0% (by HPLC, titration analysis)
N-(2-Butyl-3-(4-(3-(dibutylamino)propoxy)benzoyl)benzofuran-5-yl)methanesulphonamide hydrochloride
Formula: C₃₁H₄₅ClN₂O₅S
Peso molecolare: 593,23 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00914940
CAS Number: 141625-93-6
Merck Index: 14,03449

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 142.0 to 146.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR