3,5-Dibromoisonicotinaldehyde ≥97.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3,5-Dibromo-4-formylpyridine, 3,5-Dibromo-4-pyridinecarboxaldehyde, 3,5-Dibromoisonicotinaldehyde
D4674-1G
D4674-5G
TCIAD4674-1GEA
31
EUR
TCIAD4674-1G
TCIAD4674-5G
3,5-Dibromoisonicotinaldehyde ≥97.0% (by GC)
3,5-Dibromoisonicotinaldehyde
Formula:
C6H3Br2NO Peso molecolare: 264,9000 g/mol |
MDL Number:
MFCD06410681 CAS Number: 70201-42-2 |
Risultati del test di specificazione
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 118.0 to 122.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...