Stampa…

Diethylamine hydrobromide ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

D4667-1G D4667-5G
TCIAD4667-1GEA 16 EUR
TCIAD4667-1G TCIAD4667-5G
Diethylamine hydrobromide ≥98.0% (by titrimetric analysis)
Diethylamine hydrobromide
Formula: C₄H₁₂BrN
Peso molecolare: 154,05 g/mol
MDL Number: MFCD00012498
CAS Number: 6274-12-0
EINECS: 228-466-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Melting point 217.0 to 221.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR