2,5-Diethynylpyridine ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
D4664-250MG
D4664-1G
TCIAD4664-250MGEA
71
EUR
TCIAD4664-250MG
TCIAD4664-1G
2,5-Diethynylpyridine ≥98.0% (by GC)
2,5-Diethynylpyridine
Formula:
C₉H₅N Peso molecolare: 127,15 g/mol Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD11109672 CAS Number: 137000-75-0 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 95.0 % |
Melting point | 87.0 to 91.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...