Stampa…

1,3-Dibromoazulene ≥98.0% (by GC)

Fornitore: TCI
Attenzione

D4662-200MG D4662-1G
TCIAD4662-200MGEA 121 EUR
TCIAD4662-200MG TCIAD4662-1G
1,3-Dibromoazulene ≥98.0% (by GC)
1,3-Dibromoazulene
Formula: C₁₀H₆Br₂
CAS Number: 14658-95-8

Visualizza i codici

Risultati del test di specificazione

Appearance Dark green to Dark purple to Black powder to crystal
Purity (GC) min. 98.0 %
Melting point 88.0 to 92.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR