trans-2,6-Diisopropyl-N-(2-pyridylmethylene)aniline ≥95.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
(E)-N-(2,6-Diisopropylphenyl)-1-(pyridin-2-yl)methanimine
D4652-200MG
D4652-1G
TCIAD4652-200MGEA
108
EUR
TCIAD4652-200MG
TCIAD4652-1G
trans-2,6-Diisopropyl-N-(2-pyridylmethylene)aniline ≥95.0% (by GC)
trans-2,6-Diisopropyl-N-(2-pyridylmethylene)aniline
Formula:
C₁₈H₂₂N₂ Peso molecolare: 266,39 g/mol |
MDL Number:
MFCD06669887 CAS Number: 908294-68-8 |
Risultati del test di specificazione
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...