Diethylamine hydroiodide ≥97.0% (by total nitrogen and titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
D4643-1G
D4643-5G
TCIAD4643-1GEA
42
EUR
TCIAD4643-1G
TCIAD4643-5G
Diethylamine hydroiodide ≥97.0% (by total nitrogen and titration analysis)
Diethylamine hydroiodide
Formula:
C₄H₁₁N |
CAS Number:
19833-78-4 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 97.0 % |
Purity (Argentmetric Titration) | min. 97.0 % |
Melting point | 173.0 to 177.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...