Stampa…

2,3-Difluorobenzyl alcohol ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

D4619-5G D4619-25G
TCIAD4619-5GEA 38 EUR
TCIAD4619-5G TCIAD4619-25G
2,3-Difluorobenzyl alcohol ≥98.0% (by GC)
2,3-Difluorobenzyl alcohol
Formula: C₇H₆F₂O
Peso molecolare: 144,12 g/mol
Flash Pt: 94°C
MDL Number: MFCD00010312
CAS Number: 75853-18-8

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR