Stampa…

4,4'-Diiodo-2,2'-dimethylbiphenyl ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 4,4'-Diiodo-2,2'-dimethyl-1,1'-biphenyl

D4608-1G D4608-5G
TCIAD4608-1GEA 84 EUR
TCIAD4608-1G TCIAD4608-5G
4,4'-Diiodo-2,2'-dimethylbiphenyl ≥98.0% (by GC)
4,4'-Diiodo-2,2'-dimethylbiphenyl
Formula: C₁₄H₁₂I₂
Peso molecolare: 434,06 g/mol
MDL Number: MFCD13185944
CAS Number: 69571-02-4

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 96.0 to 100.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR