Stampa…

(2,3-Difluorophenyl)acetonitrile ≥98.0% (by GC)

Fornitore: TCI
Pericolo

Synonyms: 2,3-Difluorobenzyl cyanide, 2-(2,3-Difluorophenyl)acetonitrile, 2,3-Difluorophenylacetonitrile

D4602-1G D4602-5G
TCIAD4602-1GEA 18 EUR
TCIAD4602-1G TCIAD4602-5G
(2,3-Difluorophenyl)acetonitrile ≥98.0% (by GC)
(2,3-Difluorophenyl)acetonitrile
Formula: C₈H₅F₂N
MDL Number: MFCD00061277
CAS Number: 145689-34-5
UN: 3276
ADR: 6.1,III

Visualizza i codici

Risultati del test di specificazione

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR