Stampa…

4,6-Dichloro-5-(2-methoxyphenoxy)-2,2-bipyrimidine ≥98.0% (by GC, total nitrogen)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4,6-Dichloro-5-(2-methoxyphenoxy)-2,2'-bipyrimidine

D4598-1G D4598-5G
TCIAD4598-1GEA 28 EUR
TCIAD4598-1G TCIAD4598-5G
4,6-Dichloro-5-(2-methoxyphenoxy)-2,2-bipyrimidine ≥98.0% (by GC, total nitrogen)
4,6-Dichloro-5-(2-methoxyphenoxy)-2,2-bipyrimidine
Formula: C₁₅H₁₀Cl₂N₄O₂
Peso molecolare: 349,17 g/mol
MDL Number: MFCD03839838
CAS Number: 150728-13-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %
Melting point 157.0 to 161.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR