(2R,3S,4R,5R)-5-((Benzoyloxy)methyl)-3-fluorotetrahydrofuran-2,4-diyl dibenzoate ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
1,3,5-Tri-O-benzoyl-2-deoxy-2-fluoro-α-D-arabinofuranose, 2-Deoxy-2-fluoro-1,3,5-tri-O-benzoyl-α-D-arabinofuranose
D4594-1G
D4594-5G
TCIAD4594-1GEA
26
EUR
TCIAD4594-1G
TCIAD4594-5G
(2R,3S,4R,5R)-5-((Benzoyloxy)methyl)-3-fluorotetrahydrofuran-2,4-diyl dibenzoate ≥98.0% (by GC)
(2R,3S,4R,5R)-5-((Benzoyloxy)methyl)-3-fluorotetrahydrofuran-2,4-diyl dibenzoate
Formula:
C₂₆H₂₁FO₇ Peso molecolare: 464,44 g/mol |
MDL Number:
MFCD00083339 CAS Number: 97614-43-2 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 83.0 to 87.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...