Stampa…

Dihydrolipoic acid ≥97.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: DL-α-Lipoic Acid, Dihydro, DHLA, 6,8-Dimercapto-n-octanoic acid

D4586-200MG D4586-1G
TCIAD4586-200MGEA 134 EUR
TCIAD4586-200MG TCIAD4586-1G
Dihydrolipoic acid ≥97.0% (by GC, titration analysis)
Dihydrolipoic acid
Formula: C₈H₁₆O₂S₂
Peso molecolare: 208,33 g/mol
Punto di ebollizione: 175°C
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00166974
CAS Number: 462-20-4

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light orange to Yellow clear liquid to slightly cloudy liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Purity (Iodometric Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR