Stampa…

3-(2,4-Difluorobenzoyl)propionic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-(2,4-Difluorophenyl)-4-oxobutyric Acid, 4-(2,4-Difluorophenyl)-4-oxobutanoic acid

D4585-1G D4585-5G
TCIAD4585-1GEA 77 EUR
TCIAD4585-1G TCIAD4585-5G
3-(2,4-Difluorobenzoyl)propionic acid ≥98.0% (by GC, titration analysis)
3-(2,4-Difluorobenzoyl)propionic acid
Formula: C₁₀H₈F₂O₃
Peso molecolare: 214,17 g/mol
MDL Number: MFCD00143016
CAS Number: 110931-77-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 115.0 to 119.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR