Stampa…

(2,3-Difluoro-6-nitrophenyl)acetic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

D4583-1G D4583-5G
TCIAD4583-1GEA 51 EUR
TCIAD4583-1G TCIAD4583-5G
(2,3-Difluoro-6-nitrophenyl)acetic acid ≥98.0% (by GC, titration analysis)
(2,3-Difluoro-6-nitrophenyl)acetic acid
Formula: C₈H₅F₂NO₄
MDL Number: MFCD11053839
CAS Number: 141428-47-9

Visualizza i codici

Risultati del test di specificazione

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 154.0 to 158.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR