Stampa…

4-(2,6-Dichloro-4-pyrimidyl)morpholine ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-(2,6-Dichloropyrimidin-4-yl)morpholine, 2,4-Dichloro-6-(4-morpholino)pyrimidine

D4571-1G D4571-5G
TCIAD4571-1GEA 83 EUR
TCIAD4571-1G TCIAD4571-5G
4-(2,6-Dichloro-4-pyrimidyl)morpholine ≥98.0% (by GC)
4-(2,6-Dichloro-4-pyrimidyl)morpholine
Formula: C₈H₉Cl₂N₃O
Peso molecolare: 234,08 g/mol
MDL Number: MFCD09746259
CAS Number: 52127-83-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR