3,6-Dibromo-9-(4-bromophenyl)carbazole ≥98.0% (by HPLC, total nitrogen)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3,6-Dibromo-9-(4-bromophenyl)-9H-carbazole
D4563-200MG
D4563-1G
TCIAD4563-200MGEA
62
EUR
TCIAD4563-200MG
TCIAD4563-1G
3,6-Dibromo-9-(4-bromophenyl)carbazole ≥98.0% (by HPLC, total nitrogen)
3,6-Dibromo-9-(4-bromophenyl)carbazole
Formula:
C₁₈H₁₀Br₃N Peso molecolare: 480,00 g/mol |
MDL Number:
MFCD12024284 CAS Number: 73087-83-9 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 207.0 to 211.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...