Stampa…

3,6-Diiodocarbazole ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3,6-Diiodo-9H-carbazole

D4548-5G D4548-25G
TCIAD4548-5GEA 45 EUR
TCIAD4548-5G TCIAD4548-25G
3,6-Diiodocarbazole ≥98.0% (by GC)
3,6-Diiodocarbazole
Formula: C₁₂H₇I₂N
Peso molecolare: 419,00 g/mol
MDL Number: MFCD00450017
CAS Number: 57103-02-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 205.0 to 209.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR