3,6-Diiodo-9-phenylcarbazole ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Synonyms:
3,6-Diiodo-9-phenyl-9H-carbazole
Formula:
C₁₈H₁₁I₂N Peso molecolare: 495.10 g/mol |
MDL Number:
MFCD11521285 CAS Number: 57103-21-6 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 181.0 to 185.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...