(3S,5R)-5-(2,4-Difluorophenyl)-5-[(1H-1,2,4-triazol-1-yl)methyl]oxolan-3-ylmethyl-p-toluenesulphonate ≥98.0% (by HPLC)
Fornitore: TCI EUROPE
Synonyms:
((3R,5R)-5-((1H-1,2,4-Triazol-1-yl)methyl)-5-(2,4-difluorophenyl)tetrahydrofuran-3-yl)methyl 4-methylbenzenesulphonate, p-Toluenesulphonic acid (3S,5R)-5-(2,4-Difluorophenyl)-5-[(1H-1,2,4-triazol-1-yl)methyl]oxolan-3-ylmethyl Ester, ((3S,5R)-5-((1H-1,2,4-Triazol-1-yl)methyl)-5-(2,4-difluorophenyl)tetrahydrofuran-3-yl)methyl 4-methylbenzenesulphonate, Posaconazole Impurity 29
Formula:
C₂₁H₂₁F₂N₃O₄S Peso molecolare: 449,47 g/mol Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD13195567 CAS Number: 149809-43-8 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Elemental analysis (Nitrogen) | 8.70 to 9.70 % |
Melting point | 105.0 to 109.0 deg-C |
Specific rotation [a]20/D | -41.0 to -44.0 deg (C=1, CHCl3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...