N,N'-Dicyclohexyl-1,2-ethanediamine hydrate ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
1,2-Bis(cyclohexylamino)ethane
D4518-5G
D4518-25G
TCIAD4518-5GEA
96
EUR
TCIAD4518-5G
TCIAD4518-25G
N,N'-Dicyclohexyl-1,2-ethanediamine hydrate ≥98.0% (by GC, titration analysis)
N,N'-Dicyclohexyl-1,2-ethanediamine hydrate
Formula:
C₁₄H₂₈N₂·xH₂O Peso molecolare: 224,39 g/mol Punto di ebollizione: 312°C |
CAS Number:
4013-98-3 UN: 3259 ADR: 8,II |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % (calcd.on anh.substance) |
Water | 3.5 to 10.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...