(2,6-Difluorophenyl)acetonitrile ≥98.0% (by GC)
Fornitore: TCI
|
Pericolo
|
Synonyms:
2-(2,6-Difluorophenyl)acetonitrile, 2,6-Difluorobenzyl cyanide
Formula:
C₈H₅F₂N Punto di ebollizione: 109°C Flash Pt: 53°C |
MDL Number:
MFCD00009973 CAS Number: 654-01-3 EINECS: 211-504-7 UN: 1992 ADR: 3,III |
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...