Stampa…

2,7-Diiodofluorene ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 2,7-Diiodo-9H-fluorene

D4504-1G D4504-5G
TCIAD4504-1GEA 50 EUR
TCIAD4504-1G TCIAD4504-5G
2,7-Diiodofluorene ≥98.0% (by GC)
2,7-Diiodofluorene
Formula: C₁₃H₈I₂
Peso molecolare: 418,02 g/mol
MDL Number: MFCD00040153
CAS Number: 16218-28-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 216.0 to 220.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR