4,5-Diamino-6-hydroxypyrimidine ≥96.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
5,6-Diamino-4-hydroxypyrimidine, 5,6-Diamino-4-pyrimidinol, 5,6-Diamino-4(1H)-pyrimidinone
D4488-200MG
D4488-1G
TCIAD4488-200MGEA
49
EUR
TCIAD4488-200MG
TCIAD4488-1G
4,5-Diamino-6-hydroxypyrimidine ≥96.0% (by HPLC, titration analysis)
4,5-Diamino-6-hydroxypyrimidine
Formula:
C₄H₆N₄O Peso molecolare: 126,1200 g/mol |
MDL Number:
MFCD00006114 CAS Number: 1672-50-0 |
Risultati del test di specificazione
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 96.0 area% |
Purity (Nonaqueous Titration) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...